Learn More
BD Luer-Lok™ Caps
Catalog No. 1482331 Shop All Waters Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Syringe with Luer-lock Tip
Catalog No. For Use With (Equipment)
14-823-31 Syringe with Luer-lock Tip
1 options
This item is not returnable. View return policy
Catalog No. 14-823-31 Supplier BD Supplier No. 408531
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
For BD syringes with Luer-Lok tips
  • Female fittings
  • Sterile, single use
  • Can be used for closing off roentgenography tubing, blocking myelography needles, or blocking off stopcocks
TRUSTED_SUSTAINABILITY
Specifications
Product Type Luer-lok Cap
For Use With (Equipment) Syringe with Luer-lock Tip
Sterility Sterile