Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysing Buffer, BD

Catalog No. bdb555899 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555899 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555899 Supplier BD Biosciences Supplier No. 555899
Only null left
Add to Cart
Add to Cart

Buffered ammonium chloride-based lysing reagent

  • Concentration: 10x
  • Lyses red blood cells following monoclonal antibody staining on dilution
  • Maintains leukocytes viability after red blood cell lysis
  • Lysing solution results in good light scatter separation of lymphocytes, RBC debris by flow cytometry

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Product Type Lysis Buffer
Quantity 100mL
Validated Application RUO
Color Clear
Content And Storage 4°C
Format Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.