Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Macro-Vue™ Model 51 RPR Card Test Rotator Package

Catalog No. L4978051 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Speed Range:
100 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Speed Range
L4978051 100 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L4978051 Supplier BD Supplier No. 278051

missing translation for 'validMainframeMsg'

For use with BD Macro-Vue RPR and other card test kits

  • Rotator Package L4978051 contains one Rotator, a 5 x 7 in. Platform, and one Humidifying Cover
  • Also available is an Accessory Package L4977979 that contains one 7 x 15 in rotator platform and two humidifying covers
TRUSTED_SUSTAINABILITY

Specifications

Speed Range 100 rpm
Product Type Rotator Package
Includes 1 Humidifying cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.