Learn More
BD Macro-Vue™ RPR Cards and Components
GSA_VA
Catalog No. L4972001 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1.5 mL
10 Pack
100 Pack
300 Case
300 Pack
500 Tests
Type:
BD RPR Cards
Dispensing Needles, 20 Gauge
Liquid RPR Controls
RPR Antigen Dispensing Bottles
RPR BD Cards
RPR Control Cards
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
L4972001 100 Pack RPR BD Cards
L4970509 10 Pack RPR Antigen Dispensing Bottles
L4971849 300 Case BD RPR Cards
L4971949 300 Pack RPR BD Cards
22-130989 10 Pack Dispensing Needles, 20 Gauge
22-349688 500 Tests -
L4976909 1.5 mL Liquid RPR Controls
22-727891 10 Pack RPR Control Cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. L4972001 Supplier BD Supplier No. 272001
Add to Cart
Add to Cart
Includes
BD Macro-Vue RPR cards, 18-mm circle, quantitative, 30 spot cards
For large-scale testing in public health and clinical laboratories
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis†
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Reagin (Syphilis)
Type RPR BD Cards
For Use With (Application) For large-scale testing in public health and clinical laboratories
Includes BD Macro-Vue RPR cards, 18-mm circle, quantitative, 30 spot cards
Quantity 100 Pack