Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Macro-Vue™ RPR (Rapid Plasma Reagin) 18mm Circle Card Tests: Antigen Suspension

Catalog No. L270309B Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
3mL ampule 10/sp
3mL ampule 3/sp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L270309B 3mL ampule 10/sp
L270333B 3mL ampule 3/sp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. L270309B Supplier BD Supplier No. 270309

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.

Nontreponemal testing procedure for the serologic detection of syphilis

RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.

  • Recommended when venous blood collection employed and a large volume of serum available, such as in public health and clinical laboratories
  • Various RPR Card Test kits are available with all the necessary supplies to perform the procedure
  • A wide array of kit sizes are available
  • Antigen and testing components may be purchased separately
  • Test cards are available in 10, 15 and 30 spot cards
  • BD Dispenstirs™ are included with tests to serve as pipetting and spreading devices
  • When specimen contains antibody, flocculation occurs with coagglutination of carbon particles of RPR Card antigen, which appear as black clumps against white background of card
  • Nonreactive specimens appear to have an even light-gray color
  • In special situations when nontreponemal test results are needed rapidly and specimen is collected as EDTA plasma, RPR 18mm Circle Card Test can be used if test is performed within 24 hours

TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Reagin (Syphilis)
Type Antigen Suspension
Formulation Cardiolipin 0.003%, Lecithin 0.020 to 0.022%, Cholesterol 0.09%, EDTA 0.0125M, Na2HPO4 0.01M, KH2PO4 0.01M, Sodium Azide 0.2%, Charcoal 0.02%, Choline Chloride 10% w/v, Deionized/Distilled Water
For Use With (Application) Serologic detection of Syphilis
Quantity 3mL ampule 10/sp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.