Learn More
BD Macro-Vue™ RPR (Rapid Plasma Reagin) 18mm Circle Card Tests: Cards
Shop All BD Diagnostic Systems ProductsDescription
- Recommended when venous blood collection employed and a large volume of serum available, such as in public health and clinical laboratories
- Various RPR Card Test kits are available with all the necessary supplies to perform the procedure
- A wide array of kit sizes are available
- Antigen and testing components may be purchased separately
- Test cards are available in 10, 15 and 30 spot cards
- BD Dispenstirs™ are included with tests to serve as pipetting and spreading devices
- When specimen contains antibody, flocculation occurs with coagglutination of carbon particles of RPR Card antigen, which appear as black clumps against white background of card
- Nonreactive specimens appear to have an even light-gray color
- In special situations when nontreponemal test results are needed rapidly and specimen is collected as EDTA plasma, RPR 18mm Circle Card Test can be used if test is performed within 24 hours
Specifications
Specifications
| Detectable Analytes | Reagin (Syphilis) |
| Type | RPR Card Test Kit |
| Formulation | Cardiolipin 0.003%, Lecithin 0.020 to 0.022%, Cholesterol 0.09%, EDTA 0.0125M, Na2HPO4 0.01M, KH2PO4 0.01M, Sodium Azide 0.2%, Charcoal 0.02%, Choline Chloride 10% w/v, Deionized/Distilled Water |
| For Use With (Application) | Serologic detection of Syphilis |
| Includes | 30 Spot Cards |
| Quantity | 300 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.