Learn More
BD MAP Kinase Activation Sampler Kit
Catalog No. BDB612544 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB612544 1 Kit
1 options
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Catalog No. BDB612544 Supplier BD Supplier No. 612544
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Includes
ERK1, ERK1/2 (pT202/pY204), pan-JNK/SAPK1, JNK (pT183/pY185), p38a/SAPK2a, p38 MAPK, Storage Buffer:aqueous buffered solution containing BSA, glycerol, and ≥0.09% sodium azide
For blotting procedures
  • Format: Purified
  • Activates multiple signal transduction pathways
  • Regulates cell growth and differentiation
Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research use only