Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ BD MAX ENTERIC PARASITE CONTROL PANEL
SDP

Catalog No. 23047305 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-305 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-305 Supplier Microbiologics™ Supplier No. 8204
Add to Cart
Add to Cart
This item is not returnable. View return policy

The BD MAX™ Enteric Parasite Control Panel includes 6 inactivated microorganism pellets each comprised of the 3 organisms recommended by the manufacturer for QC listed below (6 vials/pellets total): Cryptosporidium parvum, Giardia lamblia, Recombinant Entamoeba histolytica

  • For use with the BD MAX™ Enteric Parasite Panel
  • Pellets are stable at room-temperature
  • Shipping restriction: Currently not avaliable for shipment to Canada and Europe

Specifications

Biosafety Level 1
Content And Storage 2°C to 8°C
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) For in vitro diagnostic use
Packaging Type Plastic Box
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.