Learn More
MicroBiologics™ BD Max™ Extended Enteric Bacterial Verification Panel
Shop All Microbiologics Products

Description
- Intact pathogens inactivated by chemical, radiological, or heat treatments
Organisms
- S. Typhimurium derived from NCTC 74
- S. sonnei derived from NCTC 12984
- E. coli derived from CDC 2010C-3114 (STEC)
- C. jejuni derived from NCTC 11322
- P. shigelloides derived from NCTC 10363
- Y. enterocolitica derived from NCTC 11174
- E. coli derived from NCTC 11602 (STEC)
- V. parahaemolyticus derived from NCTC 13209
Specifications
Specifications
| Content And Storage | 2 to 25°C |
| Format | Helix Elite™ single use foil pouch |
| Product Type | Lyophilized Organism |
| Target Organism Class | Enteric Bacteria |
| Test Type | Verification Panel |
| For Use With (Application) | Used as positive control |
| For Use With (Equipment) | Cepheid Xpert™ assay kits |
| Quantity | 21 Pellets of 8 Organisms |
| Specimen Type | Inactivated Lyophilized Microorganisms |
† The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.