Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MicroFlow™ Mouse Micronuclei Plus Kit

Catalog No. bdb552730 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
60 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552730 60 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552730 Supplier BD Supplier No. 552730

Includes

Anticoagulant/diluent, buffer solution, long-term storage solution (LTSS), RNase solution, mouse anti-CD71 antibody, DNA stain, platelet antibody, biological standards (positive control samples, negative control samples, CD71-negative setup samples and Malaria Biostandard blood samples), CD-ROM

For preparing mouse blood samples for flow cytometric enumeration of micronucleated erythrocyte populations

  • Reactivity: mouse
  • Size: 60 tests

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage -75° to -85°C
Includes Anticoagulant/diluent, buffer solution, long-term storage solution (LTSS), RNase solution, mouse Anti-CD71 antibody, DNA stain, platelet antibody, biological standards, CD-ROM
For Use With (Application) For preparing mouse blood samples
No. of Tests 60
Quantity 60 Tests
For Use With (Equipment) Flow Cytometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.