Learn More
BD MicroFlow™ Rat Micronuclei Analysis Plus Kit
Catalog No. BDB552731 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
60 Tests
Catalog No. Quantity
BDB552731 60 Tests
1 options
Catalog No. BDB552731 Supplier BD Supplier No. 552731
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Used when preparing rat blood samples for flow cytometric enumeration of micronucleated erythrocyte populations
MicroFlow method components are the biological standards which aid flow cytometer configuration for the micronucleus scoring application.
  • Offers advantages over other automated methods of analysis
  • Has significant advantages compared to tradition al microscopic scoring techniques
TRUSTED_SUSTAINABILITY
Specifications
Host Species Rat
Quantity 60 Tests