Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Microtainer™ Plastic Capillary Blood Collectors

Catalog No. 0266938 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
500 μL
Additive:
K2 EDTA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive
02-669-38 500 μL K2 EDTA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-669-38 Supplier BD Supplier No. 365973/DEL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for collecting, transporting and processing small quantities of blood obtained by skin puncture

  • FloTop collectors (on top of the tube) allow faster and more efficient collection
  • Color-coded stoppers simplify sample identification and additive type
  • FloCoat™ process for dispensing EDTA delivers a more uniform distribution throughout the tube, assuring a proper blood-to-additive ratio and faster anti-coagulation
  • Volume-level lines on most tubes also help ensure proper blood-to-additive ratio

TRUSTED_SUSTAINABILITY

Specifications

Product Type Whole Blood Tube
Additive K2 EDTA
Material Plastic
Closure Type Color-coded Stopper
Capacity (Metric) 500 μL
Closure Included Yes
Closure Color Lavender
For Use With (Application) Collecting, transporting and processing small quantities of blood obtained by skin puncture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.