Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Midget Urinometer

Catalog No. 1138422 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
For Use With (Application):
Urine analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
11-384-22 Urine analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-384-22 Supplier BD Supplier No. 421525
This item is not returnable. View return policy

Set requires only a small amount of urine for testing

  • Reads specific gravity from 1.000 to 1.040
  • Hydrometer has a solid shot-weighted float and an extra long stem for grasping

Order Info

Jar is 5 in. (13cm) high with extra wide base.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Urine analysis
Specific Gravity Range 1 to 1.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.