Learn More
BD Monolight™ Luminometer Cuvettes
Catalog No. BDB556862 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB556862 1 Ea.
1 options
Catalog No. BDB556862 Supplier BD Supplier No. 556862
For multi-application gel imaging
  • Plastic
  • For use with BD™ Monolight™ Luminometers
  • Size: 12 x 75mm
TRUSTED_SUSTAINABILITY
Specifications
Type Luminometer Cuvette
Material Plastic
Quantity 1 Ea.
For Use With (Equipment) Luminometer
For Use With (Application) Gel Imaging