Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse Cell Surface Marker Screening Panel

Catalog No. bdb562208 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562208 5 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Please read through instructions before you begin
Catalog No. BDB562208 Supplier BD Supplier No. 562208
Please read through instructions before you begin

Includes

Component 51-9007606AK- Mouse Cell Surface Marker Panel - Part A

  • Mouse Cell Surface Marker Lyoplate Plate 1
  • Mouse Cell Surface Marker Lyoplate Plate 2
  • Mouse Cell Surface Marker Lyoplate Plate 3
Component 51-9007606BK-Mouse Cell Surface Marker Screening Panel - Part B
  • Biotin Goat Anti-Rat Ig (1.0mL)
  • Biotin Goat Anti-Mouse Ig (0.25mL)
  • Biotin Goat Anti-Armenian Hamster Ig (0.25mL)
  • Biotin Goat Anti-Syrian Hamster Ig (0.1mL)
  • Alexa Fluor™ 647 Streptavidin (0.2mL)

For screening cell lines, primary cells or tissues

  • Contains 176 purified monoclonal antibodies that are specific to different cell surface markers
  • Includes immunoglobulin (Ig) isotype controls (rat, mouse, and hamster) for assessing nonspecific background staining
  • Three antibody-containing 96-well plates with 2.75μg of primary antibody each
  • Shows compatibility with cells that express fluorescent reporter genes, such as green fluorescent protein (GFP)
  • Panel also compatible with flow cytometry and bioimaging technology platforms
  • Can be used with additional antibodies that helps in recognizing cell surface and intracellular molecules

TRUSTED_SUSTAINABILITY

Specifications

Host Species Mouse
Quantity 5 Tests
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.