Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Foxp3 buffer set, BD

Catalog No. bdb560409 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560409 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Irritating to eyes and skin. Do not breathe vapor. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing
Catalog No. BDB560409 Supplier BD Biosciences Supplier No. 560409
Only null left
Add to Cart
Add to Cart
Irritating to eyes and skin. Do not breathe vapor. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing

Includes

Mouse Foxp3 fixation concentrate (20 x), mouse Foxp3 permeabilization concentrate (5 x)

For intracellular staining of mouse Foxp3 and surface staining of appropriate CD markers

  • Optimized for use with Foxp3 mAb clone MF23
  • Acquire at least 15,000 to 25,000 CD4 positive lymphocytes

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mouse Foxp3 Buffer set
For Use With (Application) Intracellular staining of Mouse Foxp3, surface staining of appropriate CD markers, for Foxp3 mAb clone MF23
Quantity Ea.

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.