Learn More
BD Mouse Hematopoietic Stem and Progenitor Cell Isolation Kit
Catalog No. BDB560492 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
BDB560492 100 Tests
1 options
Catalog No. BDB560492 Supplier BD Supplier No. 560492
Add to Cart
Add to Cart
Includes
Monoclonal antibodies, isotype controls and other staining tools, flow cytometry compensation beads, protocols and analysis guidelines
For high purity isolation of hematopoietic progenitor and stem cells (HPCs and HSCs) from bone marrow
To perform multicolor flow cytometric cell sorting of hematopoietic progenitor cells (HPCs) and hematopoietic stem cells (HSCs) from mouse bone marrow samples
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Formulation CD34 FITC, c-Kit PE, Sca-1 PE-Cy™7 and Lineage Cocktail APC, isotype controls, 7-AAD, BD™ CompBead compensation particles
For Use With (Application) Flow cytometry
Includes Monoclonal Antibodies, isotype controls and other staining tools, flow cytometry compensation beads, protocols and analysis guidelines
Quantity 100 Tests
Research Use Only