Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse IFN-γ (AN-18) ELISA Set

Catalog No. BDB551866 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551866 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551866 Supplier BD Supplier No. 551866
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Capture Antibody: Anti-mouse IFN-Γ monoclonal antibody (clone AN-18)
  • Detection Antibody: Biotinylated anti-mouse IFN-Γ monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant mouse IFN-Γ, lyophilized
  • Instruction / Analysis Certificate

For Immunoassay protocols

The OptEIA Set for mouse interferon-Γ (IFN-Γ), utilizing clone AN-18, contains components necessary to develop enzymelinked immunosorbent assays.

  • Reactivity: Mouse
  • For natural or recombinant mouse IFN-Γ in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse IFN-gamma (AN-18) ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse IFN-g in Serum, Plasma and cell culture supernatants
Quantity 20 Plates
Storage Requirements 2°C to 8°C

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.