Learn More
Description
Includes
Stop solution, Standard/sample diluent, TMB one-Step substrate reagent, detection antibody biotin anti-mouse IFN-gamma, mouse IFN-gamma standard, ELSIA diluent III, Enzyme concentrate streptavidin-HRP, wash concentrate, mouse IFN-g antibody coated plate
The BD OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).
- 30 kD homodimeric protein, as shown by SDS-PAGE analysis
- Utilizes a monoclonal antibody specific for mouse IFN-γ coated on a 96-well plate
Applications:
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Mouse |
| Includes | Stop solution, standard/Sample diluent, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Mouse IFN-gamma, Mouse IFN-gamma standard, ELISA diluent III, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), Mouse IFN-g Antibody coated plate |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | Quantitative determination of Mouse IFN-gamma, ELISA |
| Product Type | Mouse IFN-gamma ELISA kit II |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.