Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse IgE ELISA Set

Catalog No. BDB555248 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555248 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555248 Supplier BD Supplier No. 555248
Add to Cart
Add to Cart

Includes

Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate

An immunoassay with different sensitivity and dynamic range

To develop enzyme-linked immunosorbent assays (ELISA) for mouse IgE in serum, plasma, and cell culture supernatants

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.6 to 100ng/mL
Product Type Mouse IgE ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for Mouse IgE in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse IgE monoclonal (capture) antibody, Biotinylated anti-Mouse IgE monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized purified Mouse IgE (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.