Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse IgG2a ELISA Set

Catalog No. BDB552576 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552576 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552576 Supplier BD Supplier No. 552576
Add to Cart
Add to Cart

Sufficient f0r 20 x 96-well Plates

  • Capture antibody: anti-mouse IgG2a monoclonal antibody
  • Detection antibody: biotinylated anti-mouse IgG2a monoclonal antibody
  • Standard range: 3.1 - 200ng/mL

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 3.1 to 200ng/mL
Product Type Mouse IgG2a ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or purified Mouse IgG2a in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse IgG2a monoclonal (capture) antibody, Biotinylated anti-Mouse IgG2a monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized purified Mouse IgG2a (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.