Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Capture Antibody: anti-mouse IL-6 monoclonal antibody
- Detection Antibody: Biotinylated anti-mouse IL-6 monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant mouse IL-6, lyophilized
- Instruction/Analysis Certificate
- Contains components necessary to develop enzyme-linked immunosorbent assays (ELISA)
- For natural or recombinant mouse IL-6 in serum, plasma, and cell culture supernatants
Enzyme Linked Immunosorbent Assay
Specifications
Specifications
| Product Type | Mouse IL-6 ELISA Set |
| Sufficient For | Twenty 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse IL-6 in serum, plasma and cell culture supernatants |
| Kit Contents | anti-Mouse IL-6 monoclonal (capture) antibody, Biotinylated anti-Mouse IL-6 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-6 (standard), Streptavidin-HRP Conjugate |
| Storage Requirements | 2°C to 8°C |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?