Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse Immunoglobulin Isotyping ELISA Kit

Catalog No. BDB550487 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550487 80 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550487 Supplier BD Supplier No. 550487

missing translation for 'validMainframeMsg'

Includes

8 mouse immunoglobulin isotype-specific rat monoclonal antibodies; Horseradish peroxidase (HRP)-conjugated rat anti-mouse Ig antibody; Substrate and Stop Solutions, Coating and Blocking Buffers; Positive Control Antigen Mixture

For rapid, efficient identification of mouse immunoglobulin isotypes

The Mouse Immunoglobulin Isotyping ELISA Kit employs direct horseradish peroxidase-labeled system and assay format eliminates the need for coating plate with antigen.

  • Significant reduction in assay time without sacrificing sensitivity

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Cross Reactivity Mouse
Includes 1mL Rat Anti-Mouse IgG1, 1mL Rat Anti-Mouse IgG2a, 1mL Rat Anti-Mouse IgG2b, 1mL rat Anti-Mouse IgG3, 1mL Rat Anti-Mouse IgM, 1mL Rat Anti-Mouse IgA, 1mL Anti-Mouse Igkappa, 1mL rat Anti-Mouse Ig, lambda, 1mL HRP Rat Anti-Mouse Ig, 40mL Substrate reagent
Quantity 80 Tests
Sample Type Hybridoma Culture Supernatant
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) Rapid, efficient identification of Mouse immunoglobulin isotypes, ELISA
Product Type Mouse Immunoglobulin isotyping ELISA kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.