Learn More
Description
- Cells easily express detectable levels of intracellular IL-2, TNF and IFN-Λ
- Prepared by stimulating mouse spleen cells in the presence of a protein transport inhibitor
- Contains a measurable proportion of cytokine-producing cells Storage buffer: frozen in FBS and 10% DMSO
Intracellular staining (flow cytometry)
Specifications
Specifications
| Color | Light Brown |
| Concentration | 5 x 106 cells/mL |
| Detects | IL-2, TNF and IFN-L |
| For Use With (Application) | Intracellular staining |
| Quantity | 1 mL |
| Content And Storage | ≈5 x 106 Activated Mouse splenocytes (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL. -80°C |
| Format | Frozen |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.