Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse MCP-1 ELISA Set

Catalog No. BDB555260 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555260 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555260 Supplier BD Supplier No. 555260
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yields approximately 20 plates of 96-wells

  • Contains the components necessary to develop enzymelinked immunosorbent assays (ELISA) for natural or recombinant mouse MCP-1 in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse MCP-1 ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse MCP-1 in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse MCP-1 monoclonal (capture) antibody, anti-Mouse MCP-1 monoclonal (detection) antibody Conjugated to Horseradish Peroxidase, Instruction/analysis certificate, Lyophilized recombinant Mouse MCP-1 (standard)
Quantity 20 Plates
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.