Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse Naive/Memory T Cell Panel

Catalog No. BDB561609 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561609 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561609 Supplier BD Supplier No. 561609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Alexa Fluor™ 647 Mouse Anti-Human CD197 (CCR7), APC-H7 Mouse Anti-Human CD3, PerCP-Cy™5.5 Mouse Anti-Human CD4, FITC Mouse Anti-Human CD45RA

Contains fluorescent antibodies specific for CD44, CD62L, CD4 and CD3 cell surface antigens

  • Designed to standardize multicolor staining and flow cytometric characterization
  • Contains antibodies optimized at 5μl per test
  • Formats and concentration optimized for each specificity to work as a combined panel
  • Compatible with dual-laser flow cytometers such as the BD FACSCalibur™

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Host Species Mouse
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.