Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse Pluripotent Stem Cell Transcription Factor Analysis Kit

Catalog No. bdb560585 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560585 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560585 Supplier BD Supplier No. 560585

Includes

Monoclonal antibodies, isotype controls,staining and fixation buffers, verified protocols, and software analysis guidelines

For measuring expression of the stem cell master regulators'Nanog, Oct3/4, and Sox2'in heterogenous stem cell populations

To perform multicolor flow cytometry on mouse pluripotent stem cells such as mouse embryonic stem cells (mESCs) and induced pluripotent stem (iPS) cells

  • Reactivity: Mouse

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Formulation Nanog PE, Oct 3/4 PerCP-Cy™5.5, Sox2 Alexa Fluor™ 647, isotype control Antibodies, BD™ CompBead Plus compensation particles, BD Cytofix™ Fixation Buffer, BD Perm/Wash™
For Use With (Application) Flow cytometry
Includes Monoclonal Antibodies, Isotype controls, Staining and fixation Buffers, verified protocols and software analysis guidelines
Quantity 50 Tests

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.