Learn More
Description
The three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse T Lymphocyte Subset Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin
- Permits recognition of each of three antigens on each cell in a sample
- Levels of expression of three antigens distinguish the major subpopulations of developing and peripheral T lymphocytes
- Additional fluorochrome-labeled reagents may be combined to further characterize T-cell subpopulations
- Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography
- Clone: 145-2C11, RM4-5, 53-6.7
- Isotype: Armenian Hamster IgG1, κ, Rat IgG2a, κ
Specifications
Specifications
| Classification | Monoclonal |
| Conjugate | APC/PE/PE-Cyanine7 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Content And Storage | Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.