Learn More
BD Mouse Th17/Treg Phenotyping Kit
Catalog No. BDB560767 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560767 1 Kit
1 options
Catalog No. BDB560767 Supplier BD Supplier No. 560767
Add to Cart
Add to Cart
Includes
CD4 PerCP-Cy5.5, IL-17A PE, Foxp3 Alexa Fluor 647 antibody cocktail, Foxp3 Buffer Set, BD GolgiStop Protein Transport Inhibitor
Simultaneously view TH17 and regulatory T cells in mouse model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For research use only. Not for diagnostic or therapeutic procedures.