Learn More
Description
- Contains solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)
Applications:
ELISA
Specifications
Specifications
| Antigen | TNF |
| Conjugate | Biotin |
| Cross Reactivity | Mouse |
| Includes | Mouse TNF standard, Mouse TNF Antibody coated plate, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), stop solution, standard/Sample Diluent, Enzyme diluent, ELISA diluent II, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Mouse TNF |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Quantitative determination of Mouse TNF, ELISA |
| Product Type | Mouse TNF ELISA kit |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.