Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse TNF (Mono/Mono) ELISA Set

Catalog No. BDB555268 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555268 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Toxic if ingested, inhaled, or in contact with skin
Catalog No. BDB555268 Supplier BD Supplier No. 555268
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toxic if ingested, inhaled, or in contact with skin

Includes

Capture Antibody: Anti-mouse TNF-α , Detection Antibody: Biotinylated anti-mouse TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate, Standard: Recombinant mouse TNF-α (lyophilized)

Requires

96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, microplate reader capable of measuring absorbance at 450nm, precision pipettes, graduated cylinder, deionized or distilled water, wash bottle or automated washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, plate sealers or parafilm

Determination of mouse tumor necrosis factor

  • For natural or recombinant mouse TNF in serum, plasma, and cell culture supernatants
  • Allow flexible assay design
  • Calibrated against purified Baculovirus-expressed recombinant mouse TNF
  • Intended for use as integral unit

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 615.6 to 1000pg/mL
Product Type Mouse TNF (Mono/Mono) ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse TNF in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse TNF monoclonal (capture) antibody, Biotinylated anti-Mouse TNF monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse TNF (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.