Learn More
BD Multi-Use Nestable Sharps Collectors: Brackets
Catalog No. 19065796 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
19-065796 12 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-065796 Supplier BD Supplier No. 305485
Add to Cart
Add to Cart
This item is not returnable. View return policy
Brackets for use with 8 qt., 14 qt., and 6 gal. BD Multi-Use Nestable Sharps Collectors
For Sharps Collectors, see cat. no. 1483026
TRUSTED_SUSTAINABILITY
Specifications
Quantity 12 Pk.
Autoclavable Autoclavable
For Use With (Application) For 8 qt., 14 qt., and 6 gal. multi-use nestable sharps collectors
Material Polypropylene