Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Multi-Use One-Piece Sharps Collectors: Vented

Catalog No. 14826138 Shop All Waters Corporation Products
Change view
Click to view available options
Capacity (English):
3.3 qt.
6.9 qt.
8.2 qt.
5 gal.
Dimensions (L x W x H):
5.3 x 9.1 x 5 in.
5.3 x 9.4 x 11.5 in.
5.3 x 9.4 x 13.4 in.
7.5 x 10.5 x 18 in.
Model:
Large funnel entry
Open Top
Regular funnel entry
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W x H) Model
14-826-138 5 gal. 7.5 x 10.5 x 18 in. Open Top
14-826-137 5 gal. 7.5 x 10.5 x 18 in. Large funnel entry
14-826-126 3.3 qt. 5.3 x 9.1 x 5 in. Regular funnel entry
14-826-128 8.2 qt. 5.3 x 9.4 x 13.4 in. Regular funnel entry
14-826-127 6.9 qt. 5.3 x 9.4 x 11.5 in. Regular funnel entry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-826-138 Supplier BD Supplier No. 305491
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Preassembled with one-way funnel valve

  • Rigid polypropylene construction provides puncture resistance, strength, and durability
  • All containers are leak resistant and have attached, locking lids for secure closure and safe transport
  • Bright colors and clear labels ensure quick, easy, accurate identification to promote safe handling of hazardous materials
  • Vented caps are suitable for incineration and/or autoclaving
  • Resealable plug cap reduces evaporation and splashing

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 19 L
Closure Type Vented Hinged Cap
Capacity (English) 5 gal.
Color Red
Dimensions (L x W x H) 7.5 x 10.5 x 18 in.
Disposable Reusable
Model Open Top
Size X-Large
Vent Vented Cap
For Use With (Application) Infectious waste
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.