Learn More
BD Cell Analysis 3P CD4 Low Control, 2.5 mL
Catalog No. 50620400
Encompass
BD Multi-Check CD4 Low Control is intended as a complete process control for immunophenotyping by flow cytometry. It is a control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis on BD FACSLyric, BD FACSCanto II, BD FACSCanto, and BD FACSCalibur flow cytometers.
Catalog No. 50-620-400 Supplier BD Cell Analysis 3P Supplier No. 349703
BD Multi-Check CD4 Low Control is intended as a complete process control for immunophenotyping by flow cytometry. It is a control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis on BD FACSLyric, BD FACSCanto II, BD FACSCanto, and BD FACSCalibur flow cytometers.