Learn More
BD Cell Analysis 3P Control, 5 mL
Catalog No. 50620398
Encompass
BD Multi-Check Control is intended as a complete process control for immunophenotyping by flow cytometry. It is a control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis on BD FACSLyric, BD FACSCanto II, BD FACSCanto, and BD FACSCalibur flow cytometers.
Catalog No. 50-620-398 Supplier BD Cell Analysis 3P Supplier No. 349701
BD Multi-Check Control is intended as a complete process control for immunophenotyping by flow cytometry. It is a control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis on BD FACSLyric, BD FACSCanto II, BD FACSCanto, and BD FACSCalibur flow cytometers.