Learn More
Medchemexpress LLC BD-Oligo | 1357578-02-9 | 98.18% | 571.64 | 5 MG
SDP
Catalog No. 5000386521
Encompass_Preferred
BD-Oligo is an oligomer-specific fluorescent chemical probe. It preferentially identifies Aβ oligomer assemblies over monomers or fibrils using diversity-directed fluorescent library (DOFL) screening and computational techniques. BD-Oligo can dynamically monitor oligomer formation during Aβ peptide fibril formation as Aβ is induced to form oligomers and ultimately fibrils over time. It also exhibits blood-brain barrier permeability with the ability to stain Aβ oligomers in vivo.
  • Identifies Aβ oligomer assemblies over monomers or fibrils
  • Monitors oligomer dynamics during Aβ peptide fibril formation
  • Exhibits blood-brain barrier permeability
  • Allows staining of Aβ oligomers in vivo
Catalog No. 50-003-86521 Supplier Medchemexpress LLC Supplier No. HY1711795MG
BD-Oligo is an oligomer-specific fluorescent chemical probe. It preferentially identifies Aβ oligomer assemblies over monomers or fibrils using diversity-directed fluorescent library (DOFL) screening and computational techniques. BD-Oligo can dynamically monitor oligomer formation during Aβ peptide fibril formation as Aβ is induced to form oligomers and ultimately fibrils over time. It also exhibits blood-brain barrier permeability with the ability to stain Aβ oligomers in vivo.
  • Identifies Aβ oligomer assemblies over monomers or fibrils
  • Monitors oligomer dynamics during Aβ peptide fibril formation
  • Exhibits blood-brain barrier permeability
  • Allows staining of Aβ oligomers in vivo