Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD OptEIA™ Monkey IFN-γ ELISA Set

Catalog No. BDB551492 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551492 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551492 Supplier BD Supplier No. 551492
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For natural or recombinant Interferon-γ (IFN-γ) in non-human primate (rhesus, cynomolgus, and baboon) serum, plasma and cell culture supernatants.

Standard in this set is calibrated against purified Baculovirus-expressed recombinant human IFN-γ. One monkey IFN-γ unit (U) is defined as one picogram of recombinant human IFN-γ.

  • Kit contents: capture antibody (Anti-Monkey IFN-γ monoclonal antibody), detection antibody (Biotinylated Anti-Monkey IFN-γ monoclonal antibody), enzyme reagent (Streptavidin-horseradish peroxidase conjugate SAv-HRP), standards (Recombinant IFN-γ, lyophilized), instruction/analysis certificate
  • Size: 20 tests of 96-well format

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Product Type Monkey IFN-g ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant IFN-g in non-Human Primate (rhesus,cynomolgus and baboon) Serum, Plasma and cell culture supernatants
Kit Contents anti-Monkey IFN-g monoclonal (capture) antibody, Biotinylated anti-Monkey IFN-g monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant IFN-g (standard), Streptavidin-HRP
Quantity 20 Plates
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.