Learn More
International Medical Industries (IMI) Tamper Evident Caps for BD Oral Syringes (Box of 1000)
Catalog No. NC2030210
Encompass
Prep-Lock™ Tamper Evident Caps provide an active deterrent to diversion and misuse. Securing syringes with Tamper-Evident Caps helps to ensure that medications arrive to patients uncompromised and makes a clear statement about how seriously your operation takes drug security. Tamper Evident Caps for oral syringes utilize IMI’S “Break Away” sleeve technology, making tampering immediately evident when the colored outer sleeve is broken or missing. •Featuring a retained indicator ring for improved control of disposable components. Tamper Evident Caps for Oral Syringes are designed to each manufacturer's unique specifications, the colored top indicates compatibility • Non-Sterile • 100 Units Per Bag • 10 Bags Per Box • Yellow top compatible with BD Univia™ Oral Syringes
Catalog No. NC2030210 Supplier International Medical Industries (IMI) Supplier No. 5773
Prep-Lock™ Tamper Evident Caps provide an active deterrent to diversion and misuse. Securing syringes with Tamper-Evident Caps helps to ensure that medications arrive to patients uncompromised and makes a clear statement about how seriously your operation takes drug security. Tamper Evident Caps for oral syringes utilize IMI’S “Break Away” sleeve technology, making tampering immediately evident when the colored outer sleeve is broken or missing. •Featuring a retained indicator ring for improved control of disposable components. Tamper Evident Caps for Oral Syringes are designed to each manufacturer's unique specifications, the colored top indicates compatibility • Non-Sterile • 100 Units Per Bag • 10 Bags Per Box • Yellow top compatible with BD Univia™ Oral Syringes