Learn More
BD Organelle Sampler Kit
Catalog No. BDB612740 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB612740 1 Kit
1 options
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Catalog No. BDB612740 Supplier BD Supplier No. 612740
Add to Cart
Add to Cart
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Includes
Annexin II, BiP/GRP78, b-Catenin, Caveolin 1, Connexin-43, EEA1, GM130, Integrina2/VLA-2a, Lamp-1, MAP2B, Bcl-2, Nucleoporin p62, Paxillin, ZO-1, Storage Buffer: aqueous buffered solution containing BSA, glycerol, and ≥0.09% sodium azide
For blotting and immunoassay procedures
  • Concentration: 250μg/mL
Western Blotting, Immunoprecipitation, Immunofluorescence, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research use only