Learn More
Description
- Composed of the signal sequence, the lectin domain, the EGF-like repeat, and the first two complement-binding domains of mouse P-selectin (CD62P) fused to the Fc region (hinge, CH2 and CH3) of human IgG1
- Binds to P-selectin ligand expressed on mouse neutrophils and myeloid cell lines
Immunoprecipitation, Flow Cytometry
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | Flow Cytometry, Immunoprecipitation |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Mouse P-selectin IgG Fusion Protein |
| Purification Method | Affinity Chromatography |
| Quantity | 0.1 mg |
| Storage Requirements | -20°C |
| Cross Reactivity | Mouse |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.