Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD PE Mouse NK Cell Separation Set - DM

Catalog No. BDB558004 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
PE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB558004 PE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558004 Supplier BD Supplier No. 558004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

CD49b/pan-NK Cells PE, and BD IMag anti-PE particles 2

For positive selection or depletion of NK and NK-T cells using BD IMagnet™

  • Clone: DX5
  • Format: BD IMag-DM, PE
  • Isotype: IgM, κ
  • Reactivity: Mouse
  • Immunogen: rIL-2'propagated NK1.1+ cells from C57BL/6 mice1
  • Strain host: Lewis

Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Conjugate PE
Host Species Rat
Isotype IgM κ

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.