Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perm Buffer IV (10x), BD

Catalog No. bdb560746 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560746 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560746 Supplier BD Biosciences Supplier No. 560746
Only null left
Add to Cart
Add to Cart

For improved immunofluorescent staining and detection of certain intracellular phosphorylated proteins

BD Phosflow™ Perm Buffer IV (Perm Buffer IV) is intended for the permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells (PBMC) and lysed whole blood cells.

  • Treatment with buffer and subsequent washing with staining buffer enables access for fluorescent antibodies to stain intracellular molecules
  • Buffer preserves light scattering properties of cells
  • Providies capacity for intracellular immunofluorescent staining in preparation for Phosflow cytometric analysis

Flow Cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cell Permeabilization Buffer
Concentration 10X
For Use With (Application) Permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells and lysed whole blood cells, flow cytometry, intracellular staining
Quantity 50 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.