Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perm / Wash™ Buffer I (10x), BD

Catalog No. bdb557885 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557885 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557885 Supplier BD Biosciences Supplier No. 557885
Only null left
Add to cart
Add to cart

Intracellular staining of post-translationally modified signaling proteins

Permeabilize cells and serve as an antibody diluent and cell wash buffer

  • Storage buffer: Aqueous buffered solution containing saponin, fetal bovine serum and ≤0.09% sodium azide

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perm/Wash Buffer I
Concentration 10X
For Use With (Application) Intracellular staining of post-translationally modified signaling Proteins to permeabilize cells, to serve as an antibody diluent, cell Wash Buffer, flow cytometry, used to permeablize or wash cells and to dilute antibodies for immunofluorescent
Quantity 125 mL

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.