Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ IHC Zinc Fixative

Catalog No. BDB552658 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500mL
1000mL
Dilution:
1 X
10 X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dilution
BDB552658 500mL 10 X
BDB550523 1000mL 1 X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB552658 Supplier BD Supplier No. 552658
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An alternative to formalin fixation of tissue for paraffin embedded sections

Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.

  • Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
  • Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases

There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Clear
Content And Storage Room temperature
Dilution 10 X
Product Type Fixative
Quantity 500mL
For Use With (Application) Immunohistochemical staining
pH 7.2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.