Learn More
BD Pharmingen™ MitoStatus TMRE
Catalog No. BDB564696 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
BDB564696 25 mg
1 options
Catalog No. BDB564696 Supplier BD Supplier No. 564696
Add to Cart
Add to Cart
In cells undergoing apoptosis, oxidative stress, necrosis, and other cellular processes, the mitochondrial membrane potential (ΔΨm) can become depolarized.
In cells undergoing apoptosis, pro-apoptotic Bcl-2 family proteins cause mitochondrial outer membrane permeabilization (MOMP), resulting in the release of cytochrome C and the subsequent activation of caspase-9 and the apoptotic cascade. MOMP often correlates with the loss of inner mitochondrial membrane potential (ΔΨm), which can be detected using ΔΨm-sensitive dyes. These cationic, lipophilic dyes accumulate within the mitochondria of healthy cells, but not within mitochondria that have lost ΔΨm due to induction of apoptosis or treatment with a mitochondrial uncoupler.
BD Pharmingen™ MitoStatus TMRE (Tetramethylrhodamine ethyl ester) is a fluorescent dye that is readily sequestered by active mitochondria, allowing for flow cytometric or imaging analysis to assess for apoptosis or mitochondrial depolarization. Non-apoptotic or cells with polarized mitochondria will fluoresce red while apoptotic or cells with depolarizard mitochondria will have diminished levels of red fluorescence. MitoStatus TMRE (Tetramethylrhodamine ethyl ester) has an excitation maximum of 549nm, but is well-excited by both the blue (e.g. 488 nM) and yellow-green (e.g. 561nM) lasers. It has an emission maximum of 574nm.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mg
Storage Buffer Lyophilized powder
Test Specificity Tested in Development: Human, Mouse
Content And Storage Upon arrival, store the dye powder desiccated and protected from light at ≤ -20°C until use. The dye powder is stable for at least 12 months if stored as indicated. After reconstitution with DMSO, store the stock solution at ≤ -20°C in small aliquots. The stock solution is stable for at least 6 months if stored as indicated.