Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ Streptavidin-HRP

Catalog No. BDB550946 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550946 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550946 Supplier BD Supplier No. 550946
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prediluted for convenient use

  • Tested by immunohistochemistry
  • Storage Buffer: Aqueous buffered solution containing BSA and ProClin™ 150 as preservative

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Conjugate HRP
Quantity 50 mL
Content And Storage 4°C
Form Liquid
For Use With (Application) Immunohistochemistry
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.