Learn More
BD Pharmingen™ Transcription-Factor Buffer Set
Catalog No. BDB562574 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
BDB562574 100 Tests
1 options
5x Perm/Wash Buffer contains ≤0.09% sodium azide. Sodium azide yields highly toxic hydrazoic azid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing. 4x Fix/Perm Buffer contains diethylene glycol and formaldehyde and is harmful.
Catalog No. BDB562574 Supplier BD Supplier No. 562574
Add to Cart
Add to Cart
5x Perm/Wash Buffer contains ≤0.09% sodium azide. Sodium azide yields highly toxic hydrazoic azid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing. 4x Fix/Perm Buffer contains diethylene glycol and formaldehyde and is harmful.
Includes
Fix/Perm Buffer (4X), Diluent Buffer, Perm/Wash Buffer (5X)
Optimized for fixing and permeabilizing cells prior to immunofluorescent staining and flow cytometric analysis of cells that express specific intracytoplasmic and intranuclear proteins
This set is designed to minimize processing time, to reduce nonspecific staining, to increase resolution of positively stained cells and to significantly reduce cell loss during fixation, permeabilization and staining procedures. This is useful for flow cytometric
detection of many proteins. It supports multiwell-plate high-throughput and bulk sample analyses and applications that require overnight sample storage.
  • High sensitivity for many transcription factors including FoxP3, RORgt, Bcl-6, and T-bet
  • Compatible with many cell types, cell surface staining, and tandem dyes
  • Size: 100 tests
Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flow cytometry
Includes Fix/Perm Buffer (4X), Diluent Buffer, Perm/Wash™ Buffer (5X)
Quantity 100 Tests