Learn More
BD PhoenixSpec™ Nephelometer
Catalog No. 23025700 Shop All Waters Corporation Products
Change view
Click to view available options
Type:
Nephelometer
Catalog No. Type
23-025-700 Nephelometer
1 options
Catalog No. 23-025-700 Supplier BD Supplier No. 440910
Add to Cart
Add to Cart
Includes
Nephelometer, AC adapter, 4 AA alkaline batteries
Battery-powered, portable device designed for measuring inoculum density in McFarland units.
  • Instrument can be used for any laboratory procedure that requires inoculum density adjustments
  • Device is accurate from 0.5 to 4 McFarland units in increments of 0.1 units
  • Used with BD PhoenixSpec Calibration Kit
  • Includes: BD PhoenixSpec Nephelometer, AC adapter and 4 AA alkaline batteries (not rechargeable)
Note: The PhoenixSpec Calibrator Kit (0.25, 0.5, 1.0 and 4.0 McFarland units) and PhoenixSpec AP Calibrator Kit (0.25, 0.5, 1.0, 2.0 and 4.0 McFarland units) are available separately.
TRUSTED_SUSTAINABILITY
Specifications
Type Nephelometer
For Use With (Equipment) BD BBL™ Crystal™ identification system and the BD Phoenix™ automated identification and susceptibility testing system
Includes BD PhoenixSpec Nephelometer, AC adapter, and 4 AA alkaline batteries (not rechargeable)
For Use With (Application) Measure the turbidity of microbial suspensions equivalent to McFarland standards of 0.10 to 4.50
Accuracy Accurate from 0.5-4 McFarland units in increments of 0.1 units
Battery Type AA alkaline (not rechargeable)