Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Phosflow™ Fix Buffer I

Catalog No. bdb557870 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557870 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains formaldehyde and is a harmful solution.
Catalog No. BDB557870 Supplier BD Supplier No. 557870
Only null left
Add to Cart
Add to Cart
Contains formaldehyde and is a harmful solution.

For simultaneous fixation and permeabilization of cells prior to intracellular staining

  • Contains 250mL of a 1X buffer solution
  • Optimized for use with the BD Phosflow brand of phosphorylated intracellular signaling protein-specific antibodies

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Dilution 1 X
Product Type Fixation Buffer
Quantity 250mL
For Use With (Application) Simultaneous fixation and permeabilization of cells prior to intracellular staining
pH 7.2
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.