Learn More
BD Phosflow™ Human Monocyte/NK Cell Activation Kit
Catalog No. BDB562089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
BDB562089 50 Tests
1 options
Catalog No. BDB562089 Supplier BD Supplier No. 562089
Add to Cart
Add to Cart
Includes
Antibody cocktail, fluorochrome-labled antibodies to six key phosphoproteins, optimized protocols and buffers
Study monocyte and NK cell signaling events without prior isolation of cells from whole blood
Complete system for the determination of the activation status of six different key phosphoproteins in B cells, monocytes, NK cells, and T cells using whole blood samples.
Evaluate:
  • Phosphorylation status information on the intracellular MAPK and JAK/STAT signaling pathway
  • Responses transduced by treated human monocytes and NK cells
  • Signaling events in T and B-cell populations
Benefits:
  • Lyophilized positive and negative control cells to increase confidence and standardization
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Host Species Human
Quantity 50 Tests
Product Type Human Monocyte/NK Cell Activation Kit